Learn More
POP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12029-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Component of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5′-ends. Also a component of RNase MRP.Specifications
POP1 | |
Polyclonal | |
Unconjugated | |
POP1 | |
hPOP1, KIAA0061, POP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10940, 315045, 67724 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q99575 | |
Pop1 | |
POP1 Fusion Protein Ag2660 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |