Learn More
Ponatinib, MedChemExpress
Product Code. 16494948
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16494948
Brand: Medchem Express HY12047
MedChemExpress Ponatinib (AP24534) is an orally active multi-targeted kinase inhibitor with IC50s of 0.37 nM, 1.1 nM, 1.5 nM, 2.2 nM, and 5.4 nM for Abl, PDGFRα, VEGFR2, FGFR1, and Src, respectively.
Ponatinib (AP24534) is an orally active multi-targeted kinase inhibitor with IC50s of 0.37 nM, 1.1 nM, 1.5 nM, 2.2 nM, and 5.4 nM for Abl, PDGFRα, VEGFR2, FGFR1, and Src, respectively.
Specifications
Chemical Name or Material Ponatinib
Physical Form Solid
CAS 943319-70-8
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C29H27F3N6O
For Use With (Application) Cancer-Kinase/protease
Synonym AP24534
Solubility Information DMSO : 50 mg/mL (93.89 mM; Need ultrasonic)
SMILES CC1=C(C=C(C=C1)C(NC2=CC(C(F)(F)F)=C(C=C2)CN3CCN(CC3)C)=O)C#CC4=CN=C5N4N=CC=C5
Molecular Weight (g/mol) 532.56
Formula Weight 532.56
Percent Purity 97.56%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.