Learn More
Azlon™ POM Autoclavable Tweezer
Click to view available options
Length (Metric):
18.0 cm
25.0 cm
Unit Size:
Each
Description
Polyoxymethylene (POM) construction. Azlon™ POM Autoclavable Tweezers are suitable for multiple applications.
- High quality, autoclavable tweezers
- Rounded ends
- Fine tooth grips for secure handling of samples
Specifications
Specifications
| Tip Style | Round |
| Length (English) | 7.09 in. |
| Material | POM |
| Length (Metric) | 18.0 cm |
| Product Type | Tweezers |
| For Use With (Application) | For secure handling of samples |
| Dimensions (L x W) | 180 L mm |
| Quantity | Ea. |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur