Learn More
Contec™ Polywipe-C Wipes
GREENER_CHOICE
Product Code. 30245520 Shop All Contec Products
Click to view available options
Dimensions (L x W):
9 x 9 in.
Unit Size:
Pack of 24
Product Code. 30245520
Brand: Contec™ PC99
Only null left
Add to basket
Add to basket
Maintain cleanliness in ISO Class 5 cleanrooms. Contec™ Polywipe-C Wipers are a cost-effective upgrade from less critical wipes. A soft, nonabrasive texture cleans surfaces without scratching.
  • The lightweight Polywipe-C wipe is designed for general-purpose cleaning in ISO Class 5 (FED-STD-209E Class 100/M-3.5) environments
  • Ideal for wiping interior surfaces of process tools and other equipment
  • A cost-effective upgrade from less critical wipes
  • Made with 100% polyester interlock knit for good solvent sorbency and abrasion and chemical resistance
  • Textured surface cleans without scratching
  • Laundered and packaged in a ISO Class 4 (FED-STD-209E Class 100/M-2.5) cleanroom
  • Lightweight 100% polyester interlock knit wiper offers good sorbency with solvents
  • Ideal for cleaning interior surfaces of process tools and other equipment
  • Soft, nonabrasive texture cleans surfaces without scratching
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Wipe
Length (English) 9 in.
Length (Metric) 22.9 cm
Width (English) 9 in.
Width (Metric) 22.9 cm
Material Polyester
Certifications/Compliance For ISO Class 5 applications
Dimensions (L x W) 9 x 9 in.
For Use With (Application) Cleanroom