Learn More
mb2 Swivel Antistatic Work Chair in Polyurethane
Product Code. 13446329
Change view
Click to view available options
Height (Metric):
870 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
13446329 870 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13446329 Supplier mb2 Supplier No. 231B3C1D2E2
  • ONLINE EXCLUSIVE PRICE
    5680.00 NOK
    5615.41 NOK / Pack of 1
    Save 64.59 NOK
    1% Off
View all promotions
Estimated Shipment: 08-10-2026
Add to basket
Add to basket
Swivel antistatic work chair in polyurethane, high gas lift with footrest and castors
  • Antistatic work chair
  • Seat and backrest in multilayers of beech and poplar wood, entirely foamed with black antistatic polyurethane
  • Backrest support in chromed steel rod, in height and depth adjustable
  • Gas height adjustment
  • Antistatic double rotating self-braking castors
  • 5-spoke base in die-cast polished aluminium
Specifications
Product Type ESD Chair
Color Black
Adjustable (Height) Yes
Adjustable Height
Autoclavable Not Autoclavable
Back Style Standard
Base Style Branched 5 Wheel
Foot Type 3-Riveted Metal strapping on Metal branches
Height (Metric) 870 mm
Length (Metric) 370 mm
Quantity 1 Ea.
Seating Height 560 to 800 mm
Weight (Metric) 12.76 kg
With Armrest No
With Backrest Yes
With Casters Yes
Footrest Yes
With Lumbar Support No
With Tilt No
Material Polyurethane
Seating Type Stool with Back