Learn More
mb2 Polyurethane Chair on Glides - Asynchronous Mechanism
Product Code. 11927264
Change view
Click to view available options
:
Height: 660mm
Product Code.
11927264 Height: 660mm
1 options
Product Code. 11927264
Brand: mb2 AP3XB2D1NE3
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Polyurethane chair on glides - Mid lift - Asynchronous mechanism
  • Chair in polyurethane
  • Seat and back in polyurethane
  • Asynchronous mecanism, adjustable seat and backrest
  • Pneumatic cylinder mid
  • 5-spoke base injected in nylon and fiberglass reinforced by a galvanized steel ring.
  • glides
Specifications
Product Type Laboratory Seat
Color Black
Adjustable (Height) Yes
Adjustable Height
Autoclavable Not Autoclavable
Back Style Standard
For Use With (Equipment) For Benchtop
Height (Metric) 660 mm
Quantity 1 Ea.
Seating Height 450 to 630 mm
Weight (Metric) 11.60 kg
With Armrest No
With Backrest Yes
With Casters No
Footrest No
With Lumbar Support No
With Tilt Yes
Material Polyurethane
Seating Type Medium Bench Height