Learn More
LABPLAS Polyurethane Sani-Stick with Dey-Engley Neutralizing Broth
Description
- Polyurethane Sani-Stick
- Saturated with Dey-Engley neutralizing broth
- Dimensions: 160 x 229 mm
- Holds: 600 mL
- Option to include gloves with kit
Specifications
Specifications
| Product Type | Hygiene Sponge Sampling Kit |
| Media Type | Neutralizers |
| Sterility | Sterile |
| Includes | Gloves |