Learn More
mb2 Polyurethane Chair on Glides
Product Code. 11917834
Change view
Click to view available options
:
Height: 700mm
Product Code.
11917834 Height: 700mm
1 options
Product Code. 11917834
Brand: mb2 AP1B2D1NE3
Polyurethane chair on glides - Mid lift - Fix mechanism
  • Chair in polyurethane
  • Seat and back in polyurethane
  • Backrest adjustable in depth by wheel.
  • Pneumatic cylinder mid
  • 5-spoke base injected in nylon and fiberglass reinforced by a galvanized steel ring.
  • Glides
Specifications
Product Type Laboratory Seat
Color Black
Adjustable (Height) Yes
Adjustable Height
Autoclavable Not Autoclavable
Back Style Standard
For Use With (Equipment) For Benchtop
Height (Metric) 700 mm
Quantity 1 Ea.
Seating Height 490 to 680 mm
Weight (Metric) 7.38 kg
With Armrest No
With Backrest Yes
With Casters No
Footrest No
With Lumbar Support No
With Tilt No
Material Polyurethane
Seating Type Stool with Back