Learn More
Kinematica™ Polytron™ Homogenizer Stands: Safety Rings
Prevents homogenizer from moving out of position.
Brand:  Kinematica™ 11045035
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10002153
  • 625.00 NOK / Each
Estimated Shipment: 20-06-2024
Description
The safety ring prevents uncontrolled lowering of the drive unit. Can also be used to fixing the drive unit height.
Specifications
Specifications
Safety Positioning Ring
Stainless Steel
ST-P10/600 stand