Learn More
Honeywell™ POLYTRIL™ Black Gloves
Click to view available options
Size:
6
7
8
9
Unit Size:
1 pair
Description
- Black polyamide knitted glove
- Black oil proof nitrile coating on palm and fingertips
- The knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
- The glove offers maximum comfort and added sturdiness
- The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
- The ventilated back and PU coating optimise the glove's breathability
- The knitted and elasticated wrist gives the glove good support
- The filament yarn reduce linting for the most sensitive applications and easily laundered for extended use
- The nitrile coating at the palm and fingertips provides outstanding abrasion resistance and impermeability to oils
- Its dark color makes it more soilproof, thus ensuring a longer life
Specifications
Specifications
| Certifications/Compliance | EN 420-03 Basic standards, EN 388-16 Mechanical hazards |
| Description | 13 gauge |
| Coating Material | Polyamide, Nitrile |
| Color | Black |
| Size | 8 |
| Length (Metric) | 23 cm |