Learn More
Cole-Parmer™ CPC™ Plastic Quick-Disconnect Sealing Plug
Product Code. 11720219
Click to view available options
Material:
Polysulfone
Unit Size:
Pack of 5
Product Code. 11720219
Brand: Cole-Parmer™ 3131178
Only null left
Add to basket
Add to basket
Easy thumb-latch disconnection
Simply Press the thumb latch to disconnect coupling halves.
Specifications
Material Polysulfone
Min. Operating Temperature (Metric) -40°C
Packaging Quantity 5 Pack
Max. Operating Temperature (Metric) 148°C
Product Type Sealing Plug