missing translation for 'onlineSavingsMsg'
Learn More

Polysucrose 400, MP Biomedicals™

Product Code. p-4483029 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
This item is not returnable. View return policy

Product Code. 11443692

Brand: MP Biomedicals 0215632810

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 26873-85-8
Molecular Formula C15H27ClO12
Molecular Weight (g/mol) 434.819
MDL Number MFCD00081599
InChI Key IACFXVUNKCXYJM-AKSHDPDZSA-N
Synonym ficol 400
PubChem CID 88330506
IUPAC Name 2-(chloromethyl)oxirane;(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1C(O1)CCl.C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O

Specifications

Color White
Physical Form Powder
Quantity 10 g
Formula Weight ≈400,000
Chemical Name or Material Polysucrose 400
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.