Learn More
Greiner Bio-One Polystyrene Vented Petri Dishes
Product Code. 17230542
Click to view available options
Sterility:
Sterile
Diameter (Metric):
60 mm
Height (Metric):
15 mm
Unit Size:
Each
Pack of 600
Product Code. 17230542
Brand: Greiner Bio-One 628161
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Used for easy stacking. Greiner Bio-One Polystyrene Vented Petri Dishes is designed for improved gas exchange.
  • Vented for improved gas exchange
  • Manufactured from crystal polystyrene for optimal analysis by microscopy
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Polystyrene
Shape Round
Sterility Sterile
Type Petri Dish
No. per Case 600
Vent Yes
Dimensions (Dia. x H) 60 x 15 mm
Height (Metric) 15 mm
Diameter (Metric) 60 mm
For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.