Learn More
Kartell™ Polystyrene Petri Dishes
Description
- Disposable
- Clear Petri dish constructed of PS (Polystyrene)
- Designed for routine lab work
- Can be used in bacteriological research
- CE Marked, IVD Approved
Specifications
Specifications
| Certifications/Compliance | CE/IVD |
| Color | Clear |
| Disposable | Single-use |
| Material | Polystyrene |
| Shape | Round |
| Surface Treatment | Beta-Ray Sterilization |
| Well Count | 4 |
| For Use With (Application) | For routine lab work, bacteriological research |
| Includes | 4 Sections |
| No. per Pack | 500 |
| Show More |