Description
- Manufactured from polystyrene
- Without vent
- Pack of 1620, 108 bags of 15
Specifications
Specifications
| Color | Transparent |
| Disposable | Single-use |
| Material | Polystyrene |
| Shape | Round |
| No. per Pack | 1620 |
| Packaging | 108 bags of 15 |
| Height (Metric) | 16 mm |
| Diameter (Metric) | 55 mm |