Learn More
BRAND™ Polystyrene Feces Container with Blue Screw Cap
Shop All Brand Products
Click to view available options
Capacity (Metric):
30 mL
Unit Size:
Pack of 400
Description
- Polystyrene construction
- Includes label
- Screw cap includes integraded scoop
- Lid provides grip for sample scoop
- CE-marked according to IVD-Directive 98/79 CE
- Non-sterile
Specifications
Specifications
| Graduated | No |
| Material | Polystyrene |
| Sterility | Non-sterile |
| Capacity (Metric) | 30 mL |
| Certifications/Compliance | CE-marked according to IVD-Directive 98/79 CE. |
| Closure Color | Blue |
| Closure Material | Polystyrene |
| Label | Included |
| No. per Pack | 100/Pk. |