Learn More
Clark Scientific™ Polystyrene Anti-static Spill Trays
Description
- Made from anti-static, high impact polystyrene
- Sufficient depth to safely contain even the largest liquid spills
- Wipe clean for repeated use
Specifications
Specifications
| Material | Polystyrene |
| Type | Spill Tray |
| Packaging Quantity | 100/Pk. |
| Length (Metric) | 475 mm |
| Height (Metric) | 360 mm |