Learn More
Clark Scientific™ Polystyrene Anti-static Spill Trays
Product Code. 12948837
Change view
Click to view available options
Length (Metric):
475 mm
700 mm
Packaging Quantity:
100/Pk.
25/Pk.
Unit Size:
Pack of 100
Pack of 25
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Packaging Quantity Unit Size
12948837 700 mm 25/Pk. Pack of 25
12938837 475 mm 100/Pk. Pack of 100
12958837 700 mm 100/Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12948837 Supplier Clark Scientific™ Supplier No. AT2PK25
  • ONLINE EXCLUSIVE PRICE
    1765.00 NOK
    1599.90 NOK / Pack of 25
    Save 165.10 NOK
    9% Off
Estimated Shipment: 29-12-2026
Made from anti-static, high impact polystyrene, these spill trays are ideal in most research, scientific, laboratory and other environments.
  • Made from anti-static, high impact polystyrene
  • Sufficient depth to safely contain even the largest liquid spills
  • Wipe clean for repeated use
Specifications
Material Polystyrene
Type Spill Tray
Packaging Quantity 25/Pk.
Length (Metric) 700 mm
Height (Metric) 450 mm