Learn More
Schuett Biotec™ Polystyrene Aluminum Sample Dishes
Product Code. 10299040
Change view
Click to view available options
Height (Metric):
7 mm
Unit Size:
Pack of 80
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
10299040 7 mm Pack of 80
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10299040 Supplier Schuett Biotec™ Supplier No. 3628103
  • ONLINE EXCLUSIVE PRICE
    476.70 NOK
    342.07 NOK / Pack of 80
    Save 134.63 NOK
    28% Off
Estimated Shipment: 09-11-2026
Specifications
Bottom Shape Flat
Material Aluminum
Shape Round
Type Weighing Dish
Height (Metric) 7 mm
Diameter (Metric) 100 mm