Learn More
Poulten & Graf GmbH Polystop™ 3 piece stopper for Dropping Bottle
Product Code. 12879778
Change view
Click to view available options
Volume (Metric):
30 mL
50 mL
Unit Size:
Pack of 10
Product Code. Volume (Metric) unitSize
12879778 30 mL Pack of 10
10137761 50 mL Pack of 10
2 options
Product Code. 12879778
Brand: Poulten & Graf GmbH L188/SAV/A
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Poulten & Graf GmbH Polystop™ 3 piece stopper for Dropping Bottle includes a glass pitette, vinyl teat, and stopper.
  • Includes a strong ungraduated glass pipette
  • Includes a durable vinyl teat
  • Includes a PP-stopper
Specifications
Material Glass
Graduated No
Volume (Metric) 30 mL
Type 3 piece stopper for dropping bottles
For Use With (Equipment) Dropping Bottle