Learn More
Kautex™ Polypropylene Wide Mouth Bottles Series 303
Product Code. 11351994
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
Neck ID (Metric):
24.3 mm
30.9 mm
39.9 mm
50.8 mm
Unit Size:
Each
Pack of 10
Pack of 20
Pack of 4
Product Code. 11351994
Brand: Kautex™ 2000783855
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured with polypropylene for high chemical and thermal resistance. Kautex™ Polypropylene Wide Mouth Bottles are ideal for pastes, granulates and hot filled products.
  • Autoclavable up to 121° C
  • Graduated with blue ink for optimum visibility
Note: Before autoclaving screw off the closure and place it upside down on the bottle.
Application:
  • dispensing
  • sending samples
  • mailing
  • transport
  • storage
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 1000 mL
Color Translucent
For Use With (Application) Dispensing, Sending Samples, Mailing, Transport, Storage
Form Round
Includes Closure
Material Polypropylene
Neck ID (Metric) 50.8 mm
Replacement Closure Screw Closure PP
Quantity 1 Ea.
Graduated Yes
Height (Metric) 205 mm
Product Type Wide Mouth Bottle
Closure Type Screw Cap