Learn More
Welch™ Polypropylene Vacuum Regulator
Product Code. 11896423 Shop All Welch Ilmvac Products
Change view
Click to view available options
For Use With (Equipment):
Type MP/MPC 301, 601, 901, 1201
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11896423 Type MP/MPC 301, 601, 901, 1201 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11896423 Supplier Welch™ Supplier No. 700458
Estimated Shipment: 09-12-2026
Specifications
Depth (Metric) 92 mm
For Use With (Equipment) Type MP/MPC 301, 601, 901, 1201
Height (Metric) 110 mm
Material Polypropylene
Weight (Metric) 138 g
Width (Metric) 86 mm
Display Type Digital
Pressure Range 1000 to 1 mbar
Product Type Vacuum Regulator