Learn More
Kartell™ Urine Containers, Polystyrene
Click to view available options
Capacity (Metric):
200 mL
Unit Size:
Pack of 500
Description
- Individually wrapped aseptic containers prevent contamination of sample
- Attached label facilitates accurate data collection practices
- Permmanent molded graduations to identify collection volume
- CE marked
- IVD approved
Specifications
Specifications
| Product Type | Urine Specimen Container |
| Graduated | Yes |
| Material | Polystyrene |
| Capacity (Metric) | 200 mL |
| Certifications/Compliance | CE–IVD |
| Closure Material | Polypropylene |
| Closure Type | Screw Cap |
| Diameter (Metric) | 66 mm |
| For Use With (Application) | Collection of urine and biological specimens |
| Height (Metric) | 85.5 mm |
| Show More |