Learn More
Ratiolab™ Polypropylene Urine Beakers
Product Code. 15187073
Change view
Click to view available options
Capacity (Metric):
60 mL
125 mL
Unit Size:
Pack of 500
Pack of 600
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15187073 60 mL Pack of 600
10369182 125 mL Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15187073 Supplier Ratiolab™ Supplier No. 5921175
  • ONLINE EXCLUSIVE PRICE
    1050.00 NOK
    671.31 NOK / Pack of 600
    Save 378.69 NOK
    36% Off
View all promotions
Estimated Shipment: 15-12-2026
Collect urine specimens in these beakers. Ratiolab™ Polypropylene Urine Beakers are ideal for sampling, transport and storage of solid, pasty and liquid samples.
  • Available in polypropylene (PP) or polystyrene (PS)
  • Practical multi-purpose beakers and bottles
  • Ideal for the clinical routine
  • With red screw-cap
  • Supplied in non-sterile or sterile, individually packed
Specifications
Sterility Non-sterile
Quantity 600 Pk.
Capacity (Metric) 60 mL
Product Type Urine Beaker
Material Polypropylene
For Use With (Application) For sampling, transport and storage of solid, pasty and liquid samples
For Use With (Equipment) For sampling, transport and storage of solid, pasty and liquid samples