Learn More
Buerkle™ Polypropylene Universal Graduated Beakers
Product Code. p-8002477 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
400 mL
1000 mL
Unit Size:
Each
Attention! Make sure to pay attention to the List of Chemical Resistance!
Product Code. 17270674
Brand: Buerkle™ 71090100
Only null left
Add to basket
Add to basket
Attention! Make sure to pay attention to the List of Chemical Resistance!
Polypropylene Universal Graduated Beakers with indestructable graduation and three specially formed spouts for easy pouring of liquids.
  • Can be used as disposable
  • Manufactured from transparent polypropylene
  • Autoclavable
  • Space-saving: beakers stack in one another
TRUSTED_SUSTAINABILITY
Specifications
Color Transparent
Diameter (Metric) Base 31 mm
Diameter (Metric)Top 45 mm
Capacity (Metric) 100 mL
Material Polypropylene
For Use With (Application) Specially Formed Spouts Enable Easy Pouring of Liquids
Increments 5 mL
Quantity Each
Autoclavable Autoclavable
Disposable Single-use
Graduated Yes