Learn More
Kartell™ Plastilab™ Unequal Straight Tapered Tubing Connectors
BIOPHARMA
Product Code. p-8000638
Change view
Click to view available options
Port Size:
1.6/7.5mm
Bore Size: 1.6 to 4.6mm
Bore Size: 1.6 to 7.5mm
Bore Size: 4.6 to 7.5mm
Unit Size:
Pack of 10
Pack of 100
Product Code. Port Size unitSize
11914289 Bore Size: 1.6 to 4.6mm Pack of 10
11924289 Bore Size: 1.6 to 7.5mm Pack of 10
11934289 Bore Size: 4.6 to 7.5mm Pack of 10
12513317 1.6/7.5mm Pack of 100
4 options
Product Code. 11914289
Brand: Kartell™ 7087700
Only null left
Add to basket
Add to basket
Designed with different inlet and outlet diameters.
  • For use with flexible tubing
  • Autoclavable
Specifications
Diameter (Metric) 4-8 mm to 10-12 mm
Autoclavable Autoclavable
Material Polypropylene
Diameter (English) Outer 0.15 to 0.31 in., 0.39 to 0.47 in.
Diameter (Metric) Outer 4 to 8 mm, 10 to 12 mm
For Use With For flexible tube
Packaging Quantity 10/Pk.
Port Size Bore Size: 1.6 to 4.6mm
Shape Straight
Flexibility Flexible
Product Type Unequal Straight Tapered Tubing Connector