Learn More
Buerkle™ Tube Extension with Outer Thread
Product Code. 10055180 Shop All Bürkle Products
Change view
Click to view available options
Thread Size:
1/2 in.
3/4 in.
Width (Metric):
12.5 mm
18 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size Width (Metric) unitSize
10055180 1/2 in. 12.5 mm Each
10540784 3/4 in. 18 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10055180 Supplier Buerkle™ Supplier No. 85200012
  • ONLINE EXCLUSIVE PRICE
    52.70 NOK
    51.90 NOK / Pack of 1
    Save 0.80 NOK
    1% Off
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
Manufactured from polypropylene.
  • 152mm in length
  • Two different thread sizes available
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EU Foodstuffs
Autoclavable Autoclavable
Material Polypropylene
Overall Length (Metric) 152 mm
Thread Size 1/2 in.
Width (Metric) 12.5 mm
Product Type Tube Extension Outer Thread