Learn More
Ark Plas Products™ Polypropylene Threaded Female Luer
Product Code. 11781976
Click to view available options
Thread Size:
1/4-28 UNF
Unit Size:
Pack of 25
Product Code. 11781976
Brand: Ark Plas Products™ LMX33PP0
Only null left
Add to basket
Add to basket
Use these luers with a wide variety of medical and laboratory instruments. Ark Plas Products™ Polypropylene Threaded Female Luers feature conical tapers molded to ISO 594-1 and ISO 594-2 Standards for 6% luer tapers to ensure compatibility with all other luers molded to the same standard.
  • Natural polypropylene luer plugs
  • Feature barbs designed with no parting line for reliable, leak-free use with a broad variety of medical and laboratory instruments, including syringetips and needles or stopcocks and needles
  • Molded in virgin, lot-traceable material
  • Ark-Plas Luer fittings include a large selection of both male and female luers available in locking or slip tip variances, as well as locking rings, bulkhead luers for panel mounting, threaded luers, adapters and plugs
Specifications
Autoclavable Autoclavable
For Use With Flexible tubing or threaded ports
Min. Operating Temperature (Metric) -17°C
Packaging Quantity 25/Pk.
Port Size 1/4-28 UNF
Thread Size 1/4-28 UNF
Color Natural
Max. Operating Temperature (Metric) 135°C