Learn More
Buerkle™ Polypropylene Thread Adapter
Product Code. p-8003082 Shop All Bürkle Products
Click to view available options
Thread Size:
External: 2 in. BSP - 64 mm (BSI)
External: 2 in. Coarse Mauser - 64 mm (BSI)
Internal: 35 mm - 2 in. (BSP)
Unit Size:
Each
Product Code. 10014990
Brand: Buerkle™ 06003004
Only null left
Add to basket
Add to basket
Buerkle™ Polypropylene Thread Adapter facilitates the secure connection of pumps and discharge cocks to a variety of containers.
  • Constructed of polypropylene
  • Interchange the connections to different adapters
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
For Use With Pump-it Container Pump
Thread Size External: 2 in. Coarse Mauser - 64 mm (BSI)
Color Green
Product Type Thread Adapter