Learn More
Buerkle™ Polypropylene Thread Adapter
Shop All Bürkle Products
Click to view available options
Thread Size:
External: 2 in. BSP - 64 mm (BSI)
External: 2 in. Coarse Mauser - 64 mm (BSI)
Internal: 35 mm - 2 in. (BSP)
Unit Size:
Each
Description
- Constructed of polypropylene
- Interchange the connections to different adapters
Specifications
Specifications
| Material | Polypropylene |
| For Use With | Pumps |
| Thread Size | Internal: 35 mm - 2 in. (BSP) |
| Color | Black |
| Product Type | Thread Adapter |