Learn More
BRAND™ Translucent Polypropylene Straight Tubing Connectors
Product Code. p-8000583 Shop All Brand Products
Change view
Click to view available options
Overall Length (Metric):
47 mm
55 mm
57 mm
Unit Size:
Pack of 10
Product Code. Overall Length (Metric) unitSize
11833970 55 mm Pack of 10
11894140 47 mm Pack of 10
10737391 57 mm Pack of 10
3 options
Product Code. 11833970
Brand: BRAND™ 152710
Only null left
Add to basket
Add to basket
Manufactured from translucent polypropylene. BRAND™ Translucent Polypropylene Straight Tubing Connectors are ridged to fit a variety of tubing diameters.
  • Manufactured from translucent polypropylene
  • Ridged to fit a variety of tubing diameters
  • Straight shape
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Material Polypropylene
Overall Length (Metric) 55 mm
For Use With 6 to 10mm tubing I.D.
Packaging Quantity 10 Pack
Shape Straight
Color Translucent
Product Type Straight Tubing Connector