Learn More
Lenz Laborglasinstrumente™ Plastic Straight Hose Connector
Click to view available options
Thread Size:
GL14
GL18
Unit Size:
Each
Pack of 10
Description
- Autoclavable
- Available in two different thread sizes
- Silicone sealing
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| For Use With | For screw caps with bore |
| Packaging Quantity | 10 Pack |
| Thread Size | GL14 |
| Shape | Straight |
| Product Type | Straight Hose Connection |