Learn More
Lenz Laborglasinstrumente™ Plastic Straight Hose Connector
Product Code. 10070211
Click to view available options
Thread Size:
GL14
GL18
Unit Size:
Each
Pack of 10
Product Code. 10070211
Brand: Lenz Laborglasinstrumente™ 01350000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For connecting screw caps with bore. Lenz Laborglasinstrumente™ Plastic Straight Hose Connectors are made from polypropylene.
  • Autoclavable
  • Available in two different thread sizes
  • Silicone sealing
Specifications
Autoclavable Autoclavable
Material Polypropylene
For Use With For screw caps with bore
Packaging Quantity 10 Pack
Thread Size GL14
Shape Straight
Product Type Straight Hose Connection