Learn More
Azlon™ Polypropylene Straight Connector for Tubing
Description
- Straight pattern tubing connectors in white polypropylene
- For connecting tubing of similar or different sizes
- Tapered arms are rigid with steps to give firm grip
- Autoclavable at 121°C with good chemical resistance
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| For Use With | 10 to 15mm I.D. tubing |
| Shape | Straight |
| Color | White |
| Max. Operating Temperature (Metric) | 121°C |
| Product Type | Connector |