Learn More
Kartell™ Polypropylene Stirring Paddles
Constructed of polypropylene and steel for strength, durability and chemical resistance. Kartel™ Polypropylene Stirring Paddles provide a sturdy variety of paddles to fit any stirring need.
Brand:  Kartell™ 0043200
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10294471
  • 1290.00 NOK / Pack of 5
Estimated Shipment: 28-06-2024
Description
  • Steel rod covered with polypropylene provides a strong connection to the stirrer
  • Steel rods: 8 dia. x 350mmL fits standard overhead stirrer
  • Polypropylene blades are premanently fixed to the rods for stability
    • Fixed propeller for vortex stirring
    • U shape for slower but more vigorous agitation
    • Two sizes of swivel impellers enable the blades to be introduced through a small aperture then fully extend for stirring
Specifications
Specifications
Collapsible paddle, fully extends during stirring
98.5 mm W
Stirring Paddle
98.5 mm