Learn More
Buerkle™ Polypropylene Stirring Paddle
Product Code. p-7084551 Shop All Bürkle Products
Click to view available options
Length (Metric):
Total Length: 1.71 m
Total length: 1.17 m
Unit Size:
Each
Product Code. 10278864
Brand: Buerkle™ 82032100
Only null left
Add to basket
Add to basket
Stirring paddle made of polypropylene for gentle stirring in plastic and glass containers.
Stirring paddle made of polypropylene for gentle stirring in plastic and glass containers.
  • Polypropylene
  • Shatterproof
  • Warp resistant
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
Color Grey
Description Total Length 117 cm
For Use With (Application) For Gentle Stirring in Plastic and Glass Containers
Length (English) 6.1 in.
Weight (Metric) 0.24 kg
Width (Metric) 130 mm
Diameter (Metric) 20 mm (Rod Diameter)
Length (Metric) Total length: 1.17 m
Material Polypropylene
Quantity 1/Ea.
Type Stirring Paddle