Learn More
Buerkle™ Polypropylene Stirring Paddle
Product Code. 11981531 Shop All Bürkle Products
Change view
Click to view available options
Length (Metric):
Total Length: 1.71 m
Total length: 1.17 m
Unit Size:
Each
Product Code. Length (Metric) unitSize
11981531 Total Length: 1.71 m Each
10278864 Total length: 1.17 m Each
2 options
Product Code. 11981531
Brand: Buerkle™ 82033150
Only null left
Add to basket
Add to basket
Stirring paddle made of polypropylene for gentle stirring in plastic and glass containers.
Stirring paddle made of polypropylene for gentle stirring in plastic and glass containers.
  • Polypropylene
  • Shatterproof
  • Warp resistant
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
Color Grey
Description Total Length 171 cm
For Use With (Application) For Gentle Stirring in Plastic and Glass Containers
Width (Metric) 167 mm
Diameter (Metric) 20 mm (Rod Diameter)
Length (Metric) Total Length: 1.71 m
Material Polypropylene
Quantity 1/Ea.
Type Stirring Paddle