Learn More
Ismatec™ Standard Tubing Connectors, Plastic
Product Code. 10482082
Change view
Click to view available options
Tubing Size:
1.5mm O.D.
2.5mm O.D.
3.0mm O.D.
4.0mm O.D.
5.0mm O.D.
6.0mm O.D.
8.0mm I.D.
Shape:
Straight
T-shape
Y-shape
Unit Size:
Pack of 10
Product Code. Tubing Size Shape unitSize
10482082 4.0mm O.D. Straight Pack of 10
10710325 6.0mm O.D. Straight Pack of 10
10008451 1.5mm O.D. T-shape Pack of 10
10473232 6.0mm O.D. Y-shape Pack of 10
10482272 5.0mm O.D. Straight Pack of 10
10422852 2.5mm O.D. T-shape Pack of 10
10505373 3.0mm O.D. T-shape Pack of 10
10296752 8.0mm I.D. Straight Pack of 10
8 options
Product Code. 10482082
Brand: Ismatec™ ISM559A
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Variously sized and shaped tubing connectors. Ismatec™ Standard Tubing Connectors, Plastic are available in different plastic compositions.
  • Standard, non-reducing.
  • Designed for Ismatec™ pump tubing.
  • Available in different sizes, shapes, and compositions.
Specifications
Material Acetal Resin (POM, Polyoxymethylene)
For Use With For 4.0mm I.D. Tubing
Packaging Quantity 10/Pk.
Tubing Size 4.0mm O.D.
Shape Straight
Product Type Standard Tubing Connector