Learn More
SCAT Capillary cutter
Product Code. 10180090 Shop All SCAT Europe Products
Change view
Click to view available options
Color:
Blue
Unit Size:
Each
Product Code. Color unitSize
10180090 Blue Each
1 options
Product Code. 10180090
Brand: SCAT 900103
Only null left
Add to basket
Add to basket
Capillary cutter incl. spare blade
This cutter is made of PP, blue, with stainless steel blade.
Specifications
Product Type Tubing Cutter Cutting Wheel
For Use With (Equipment) HPLC Capillaries
Material Polypropylene
Color Blue
Description Clean Cut tubing cutter incl. spare blade
For Use With (Application) PFA and PP Fittings
Includes Replacement Blade
No. per Pack 1 Ea.