Learn More
Brand™ Polypropylene Stackable Tip Box
Product Code. 15134997 Shop All Brand Products
Change view
Click to view available options
Holds:
Pipette Tips (1000 μL)
Pipette Tips (200 μL)
Pipette Tips (300 μL)
Pipette Tips (50 μL)
Color:
Blue
Gray
Green
Yellow
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Color Unit Size
15134997 Pipette Tips (200 μL) Yellow Each
15144997 Pipette Tips (1000 μL) Blue Each
15124997 Pipette Tips (50 μL) Gray Each
15154997 Pipette Tips (300 μL) Green Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15134997 Supplier Brand™ Supplier No. 732992
  • ONLINE EXCLUSIVE PRICE
    72.05 NOK
    56.03 NOK / Pack of 1
    Save 16.02 NOK
    22% Off
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
Bulk Quote Request
Includes tip tray. BRAND™ Polypropylene Stackable Tip Box comes empty. Filter and pipet tips sold separately.
  • Manufactured from polypropylene
  • Includes tip tray
  • Empty, for filter and pipet tips
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow
Holds Pipette Tips (200 μL)
Material Polypropylene
Stackable Yes
For Use With (Equipment) Pipet and Filter Tips
No. per Pack 1 Pack
Quantity 1 Pack
Product Type Empty Tip Box