Learn More
Vitlab™ Polypropylene Stackable Beaker
Product Code. 10513091
Click to view available options
Capacity (Metric):
2000 mL
3000 mL
Unit Size:
Each
Pack of 12
Product Code. 10513091
Brand: Vitlab™ 484941
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these stackable beakers for a variety of general lab applications. VITLAB™ Polypropylene Stackable Beakers feature a stable handle and easily readable, printed black scale on both sides. Volume is equally visible for left and right handers.
  • Highly transparent
  • To preserve markings, cleaning at no higher than 60°C is recommended
  • Suitable for foodstuffs
Specifications
Diameter (Metric) Outer 170 mm
Capacity (Metric) 3000 mL
Material Polypropylene
Height (Metric) 242 mm