Learn More
Vitlab™ Polypropylene Spray Bottles
Product Code. 12674970
Change view
Click to view available options
Capacity (Metric):
400 mL
850 mL
Unit Size:
Each
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12674970 400 mL Pack of 5
10302611 400 mL Each
10463291 850 mL Each
15252509 850 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12674970 Supplier Vitlab™ Supplier No. 53510
  • ONLINE EXCLUSIVE PRICE
    572.00 NOK
    487.54 NOK / Pack of 5
    Save 84.46 NOK
    14% Off
View all promotions
In Stock
Lägg i varukorgen
Edge
Lägg i varukorgen
Use these bottles for spraying detergents or disinfectants, especially into difficultly accessible areas, as well as applications in thin layer chromatography. VITLAB™ Polypropylene Spray Bottles feature a sprayer insert with stable, smoothly operated pump trigger and adjustable spray nozzle.
  • White polypropylene spray bottles
  • Spray nozzle can be regulated from the finest mist (nebulizing) to a precise liquid jet
  • Range: approx. 3 to 4 meters
TRUSTED_SUSTAINABILITY
Specifikationer
Capacity (Metric) 400 mL
Color White
For Use With (Application) Ideal for spraying detergents or disinfectants, especially into difficultly accessible areas, as well as applications in thin layer chromatography
Material Polypropylene
No. per Pack 5 Pack
Closure Color White
Closure Included Yes
Closure Material Polypropylene
Closure Type Screw Cap
Autoclavable Not Autoclavable
Product Type Spray Bottle
Visa mer Visa mindre
For research purposes only
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.