Learn More
Vitlab™ Polypropylene Spray Bottles
Product Code. 10463291
Click to view available options
Capacity (Metric):
400 mL
850 mL
Unit Size:
Each
Pack of 5
Product Code. 10463291
Brand: Vitlab™ 53610
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these bottles for spraying detergents or disinfectants, especially into difficultly accessible areas, as well as applications in thin layer chromatography. VITLAB™ Polypropylene Spray Bottles feature a sprayer insert with stable, smoothly operated pump trigger and adjustable spray nozzle.
  • White polypropylene spray bottles
  • Spray nozzle can be regulated from the finest mist (nebulizing) to a precise liquid jet
  • Range: approx. 3 to 4 meters
Specifications
Capacity (Metric) 850 mL
Color White
For Use With (Application) Ideal for spraying detergents or disinfectants, especially into difficultly accessible areas, as well as applications in thin layer chromatography
Material Polypropylene
No. per Pack 5 Pack
Closure Color White
Closure Included Yes
Product Type Spray Bottle
Closure Material Polypropylene
Closure Type Screw Cap
Autoclavable Not Autoclavable
Show More Show Less
For research purposes only