Learn More
Buerkle™ 2-Part Shovel for foodstuffs
Product Code. 10521142 Shop All Bürkle Products
Change view
Click to view available options
Material:
Polypropylene
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
10521142 Polypropylene Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10521142 Supplier Buerkle™ Supplier No. 83002010
  • ONLINE EXCLUSIVE PRICE
    837.00 NOK
    821.55 NOK / Pack of 1
    Save 15.45 NOK
    1% Off
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
Polypropylene's easy-to-clean, smooth and pore-free surface makes these scoops extremely suitable for the food sector. The scoops can be also used for boiling products.
  • For foodstuffs (FDA and (EU) Nr. 10/2011)
  • Easy-to-clean, smooth and pore-free surface
  • The scoops can be also used for boiling products
TRUSTED_SUSTAINABILITY
Specifications
Product Type Two-Part Shovel
Material Polypropylene
Depth (Metric) 315 mm
Includes Scraper With Fixed Handle, Hand Shovel, Filling Scoop For Ideal For Sampling, Scoop Two-Piece With D-grip, Two-Piece Shovel With T-Handle, Shovel In One Piece For Reduced Contamination
Length (Metric) 966 mm
Width (Metric) 247 mm