Description
- Rounded corners for easy cleaning
- Made from rigid polypropylene
- Looped handle for hanging
- Holds approximately 2 liters (one half gallon)
Capacity/volumes are estimated and are not meant for taking exact sample weights/volumes. Stated volumes represent typical heaping samples.
Specifications
Specifications
| Capacity (Metric) | 22 mL |
| Product Type | Scoop |
| Width (Metric) | 32 mm |
| Dimensions (L x W) | 94 x 32 x 22 mm |
| Length (Metric) Handle | 32 mm |
| Material | Polypropylene |
| Length (Metric) | 94 mm |