Learn More
Bel-Art™ Scienceware™ Polypropylene Scoop
Product Code. 11756545 Shop All SP Scienceware Products
Click to view available options
Capacity (Metric):
22 mL
250 mL
2 L
Length (Metric) Handle:
32 mm
56 mm
122 mm
Dimensions (L x W):
94 x 32 x 22 mm
191 x 54 x 49 mm
349 x 148 x 89 mm
Unit Size:
Pack of 12
Pack of 6
Product Code. 11756545
Brand: Bel-Art™ Scienceware™ F367530000
Only null left
Add to basket
Add to basket
Designed for retrieving samples from a larger source. Bel Art™ Polypropylene Scoop is excellent for scooping anything from bulk powder to ice.
  • Rounded corners for easy cleaning
  • Made from rigid polypropylene
  • Looped handle for hanging
  • Holds approximately 2 liters (one half gallon)
Capacity/volumes are estimated and are not meant for taking exact sample weights/volumes. Stated volumes represent typical heaping samples.
Specifications
Capacity (Metric) 250 mL
Product Type Scoop
Width (Metric) 49 mm
Dimensions (L x W) 191 x 54 x 49 mm
Length (Metric) Handle 56 mm
Material Polypropylene
Length (Metric) 191 mm