Learn More
Vitlab™ Polypropylene Scoop
Product Code. 10608512
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Length (Metric):
100 mm
135 mm
160 mm
200 mm
260 mm
315 mm
385 mm
Unit Size:
Each
Pack of 12
Pack of 6
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Length (Metric) unitSize
10608512 500 mL 315 mm Pack of 6
10190111 1000 mL 385 mm Each
10769082 1000 mL 385 mm Pack of 6
10717391 100 mL 200 mm Each
10159641 100 mL 200 mm Pack of 12
10008360 10 mL 100 mm Each
11807930 10 mL 100 mm Pack of 12
10139930 250 mL 260 mm Each
12623566 250 mL 260 mm Pack of 6
10221821 500 mL 315 mm Each
10538701 25 mL 135 mm Each
10296622 25 mL 135 mm Pack of 12
10717201 50 mL 160 mm Each
12613566 50 mL 160 mm Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 10608512 Supplier Vitlab™ Supplier No. 39894
Spotlight Offers
Use promo code 28036
View all promotions
In Stock
Add to basket
Edge
Add to basket
Use these corrosion resistant scoops for applications in the food industry. VITLAB™ Polypropylene Scoops are lighter than metal, don’t dent and feature good chemical resistance. Also suitable as weighing scoops.
  • White polypropylene scoop
  • With precision formed filling edge and comfortable, stable handle
  • Easily readable volume quantities on the upper side of the handle
Specifications
Capacity (Metric) 500 mL
Color White
Product Type Measuring Scoop
For Use With (Application) For contact with food
Material Polypropylene
Length (Metric) 315 mm