Learn More
Vitlab™ Polypropylene Scoop
Description
- White polypropylene scoop
- With precision formed filling edge and comfortable, stable handle
- Easily readable volume quantities on the upper side of the handle
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Color | White |
| Product Type | Measuring Scoop |
| For Use With (Application) | For contact with food |
| Material | Polypropylene |
| Length (Metric) | 385 mm |