Description
- Dual graduations in mL and oz.
- Manufactured of translucent polypropylene
- Lids fit tightly, sold separately
Specifications
Specifications
| Quantity | 500/Pk. |
| Capacity (Metric) | 130 mL |
| Product Type | Sample Cup |
| Material | Polypropylene |
| Height (Metric) | 6.5 cm |