Learn More
Buerkle™ Polypropylene Rods for Draining Racks
Click to view available options
Product Type:
Rod
Rod Extension
Size:
Dia. x L: 10 x 100 mm
Dia. x L: 12 x 150 mm
Dia. x L: 6 x 60 mm
Color:
Blue
Transparent
Unit Size:
Each
Description
- The rods can be replaced and have their positions can be changed
- The extension can only be used with the rods of 100 mm length
Specifications
Specifications
| Product Type | Rod |
| Color | Blue |
| Material | Polypropylene |
| For Use With (Equipment) | PVC Draining Rack |
| No. per Pack | 1 |
| Size | Dia. x L: 10 x 100 mm |
| Length (Metric) | 100 mm |
| Diameter (Metric) Outer | 10 mm |